Anti-NK1.1 Mouse Monoclonal Antibody
NOVUNB100-77528
- Antibody type:Primary
- Antigen name:NK-1.1 Antigen
- Antigen symbol:NK1.1
- Clonality:Monoclonal
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Antigen synonyms:NK-RP1|Nk1|AI462337|NKR-P1.9|ly-55c,CD161 antigen-like family member C|natural killer cell-associated antigen 1|NKR-P1 40|locus C|natural killer cell receptor-P1|Ly-59|Nk-1.2|lymphocyte antigen 55c|NK1.1|Ly55c|lymphocyte antigen 55 complex|Ly59|lymphocyte antigen 59|Nk-1|Nkrp1c|natural killer cell receptor protein NKR-P1C|NKRP140|natural killer cell surface protein P1-40|killer cell lectin-like receptor subfamily A member 1C|CD161|Nk1.2|NKRP1
- Size:500 μg
- Pk:0,5 mg
Mouse Monoclonal NK1.1 Antibody (PK136). Tested Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Mouse.
Type: Primary
Antigen: NK1.1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse