Order Entry
Netherlands
ContactUsLinkComponent
Anti-CADM1 Rabbit Polyclonal Antibody
Anti-CADM1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-CADM1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB300-186
  • NOVUNB300-186SS
  • Antigen symbol:
  • CADM1
  • CADM1
  • Antigen name:
  • Cell adhesion molecule 1
  • Cell adhesion molecule 1
  • Antigen synonyms:
  • synCAM1|IGSF4|sgIGSF|SYNCAM|BL2|IGSF4A|TSLC1|NECL2|ST17|Necl-2|sTSLC-1|RA175
  • synCAM1|IGSF4|sgIGSF|SYNCAM|BL2|IGSF4A|TSLC1|NECL2|ST17|Necl-2|sTSLC-1|RA175
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 23705
  • 23705
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together