About this item
Recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor for pathogenic Neisseria. This MAb can be applied to test complement activation in pseudo-allergic reactions to acetylsalicylic acid and to test for measles virus infection of cells.
Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: 197-2B1
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC040931-500
- BNC040931-100
- antigeen symbool:
- CD46
- CD46
- Antigeen synoniemen:
- Trophoblast leucocyte common antigen|Membrane cofactor protein|Complement membrane cofactor protein|MIC10|MCP|AHUS2|TLX|Trophoblast lymphocyte cross reactive antigen|Measles virus receptor
- Trophoblast leucocyte common antigen|Membrane cofactor protein|Complement membrane cofactor protein|MIC10|MCP|AHUS2|TLX|Trophoblast lymphocyte cross reactive antigen|Measles virus receptor
- Type antilichaam:
- Primair
- Primair
- Clonality:
- Monoklonaal
- Monoklonaal
- Conjugatie:
- CF® 405S
- CF® 405S
- Clone:
- Clone 197-2B1
- Clone 197-2B1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gen-ID:
- 510402
- 510402
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescentie:
- Yes
- Yes
- Concentratie:
- 0,1 mg/ml
- 0,1 mg/ml
- Opslagbuffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide