Order Entry
Netherlands
ContactUsLinkComponent
Anti-ECM1 Mouse Monoclonal Antibody (CF405S) [clone: ECM1/792]
Anti-ECM1 Mouse Monoclonal Antibody (CF405S) [clone: ECM1/792]
Supplier:  Biotium
Certificates
Anti-ECM1 Mouse Monoclonal Antibody (CF405S) [clone: ECM1/792]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040792-100
  • BNC040792-500
  • Antigen symbol:
  • ECM1
  • ECM1
  • Antigen name:
  • IgA Secretory Component
  • IgA Secretory Component
  • Antigen synonyms:
  • URBWD|Extracellular Matrix Protein 1|ECM1|Secretory Component p85
  • URBWD|Extracellular Matrix Protein 1|ECM1|Secretory Component p85
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone ECM1/792
  • Clone ECM1/792
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 81071
  • 81071
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together