Actin is a major component of the cytoskeleton and is present in most cell types. It is highly specific to actin from smooth muscles. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actin and anti-smooth muscle actin and are negative with anti-myogenin.
Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: ACTA2/791
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat
- Catalog No:
- BNC880791-100
- BNC880791-500
- Antigen symbol:
- ACTA2
- ACTA2
- Antigen name:
- Smooth Muscle Actin
- Smooth Muscle Actin
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone ACTA2/791
- Clone ACTA2/791
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Mouse
- Mouse
- Gene ID:
- 500483
- 500483
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide