About this item
Recognizes a phosphor-protein of 45 kDa, identified as MyoD1. This MAb does not cross react with myogenin, Myf5, or Myf6. Antibody to MyoD1 labels the nuclei of myoblasts in developing muscle tissues. MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occasionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm s tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing s sarcomas and alveolar soft part sarcomas.
Type: Primary
Antigen: MYOD1
Clonality: Monoclonal
Clone: 5.8A MYD712
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Chicken, Human, Mouse, Rat
Specifications
- Catalog No:
- BNC940713-100
- BNC940713-500
- Antigen symbol:
- MYOD1
- MYOD1
- Antigen synonyms:
- Myogenic differentiation 1|Class C basic helix-loop-helix protein 1|PUM|Myogenin D1|Myogenic factor 3 (Myf-3)|Myoblast determination protein 1|bHLHc1
- Myogenic differentiation 1|Class C basic helix-loop-helix protein 1|PUM|Myogenin D1|Myogenic factor 3 (Myf-3)|Myoblast determination protein 1|bHLHc1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone 5.8A MYD712
- Clone 5.8A MYD712
- Reactivity:
- Chicken, Human, Rat, Mouse
- Chicken, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 181768
- 181768
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide