- Antibody type:Primary
- Antigen symbol:CD19
- Clonality:Monoclonal
- Clone:Clone CVID3/429
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Chimpanzee,Human
- Gene ID:652262
- Antigen synonyms:CVID3|Lymphocyte Surface Antigen|Leu-12|B-lymphocyte antigen CD19|T-cell surface antigen Leu-12|B-lymphocyte surface antigen B4
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-cell development and through terminal differentiation of B-cells into plasma cells. Anti-CD19 positivity is seen in the vast majority of B-cell neoplasms commonly at a lower intensity than normal B-cell counterparts. Plasma cell neoplasms are nearly always negative, as are T-cell neoplasms.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: CVID3/429
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human, Monkey