Order Entry
Netherlands
ContactUsLinkComponent
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: Ks19.1]
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: Ks19.1]
Supplier:  Biotium
Certificates
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: Ks19.1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0394-500
  • BNCB0394-100
  • Antigen symbol:
  • KRT19
  • KRT19
  • Antigen name:
  • Cytokeratin 19
  • Cytokeratin 19
  • Antigen synonyms:
  • k19|k1cs|krt19|Keratin 19 Keratin Type i 40kD
  • k19|k1cs|krt19|Keratin 19 Keratin Type i 40kD
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone Ks19.1
  • Clone Ks19.1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 654568
  • 654568
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together