Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD7 Mouse Monoclonal Antibody (CF568) [clone: B-F12]
Anti-CD7 Mouse Monoclonal Antibody (CF568) [clone: B-F12]
Supplier:  Biotium
Certificates
Anti-CD7 Mouse Monoclonal Antibody (CF568) [clone: B-F12]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680310-100
  • BNC680310-500
  • Antigen symbol:
  • CD7
  • CD7
  • Antigen synonyms:
  • TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • TP41|Leu9|T-cell surface antigen Leu-9|T-cell leukemia antigen|Tp40|p41|GP40
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone B-F12
  • Clone B-F12
  • Reactivity:
  • Pig, Human, Guinea pig
  • Pig, Human, Guinea pig
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 186820
  • 186820
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together