Order Entry
Netherlands
ContactUsLinkComponent
Anti-ERBB2 Mouse Monoclonal Antibody (Purified) [clone: HRB2/258]
Anti-ERBB2 Mouse Monoclonal Antibody (Purified) [clone: HRB2/258]
Supplier:  Biotium
Certificates
Anti-ERBB2 Mouse Monoclonal Antibody (Purified) [clone: HRB2/258]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0258-500
  • BNUB0258-100
  • Antigen symbol:
  • ERBB2
  • ERBB2
  • Antigen name:
  • HER-2 / CD340
  • HER-2 / CD340
  • Antigen synonyms:
  • Verb b2 Erythroblastic Leukemia Viral Oncogene Homolog 2|Neuro/Glioblastoma Derived Oncogene Homolog|CD340|p185
  • Verb b2 Erythroblastic Leukemia Viral Oncogene Homolog 2|Neuro/Glioblastoma Derived Oncogene Homolog|CD340|p185
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone HRB2/258
  • Clone HRB2/258
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 446352
  • 446352
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together