Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-ICAM3 Mouse Monoclonal Antibody (CF640R) [clone: 101-1D2]
 :  Biotium
 
Anti-ICAM3 Mouse Monoclonal Antibody (CF640R) [clone: 101-1D2]
 :  Biotium

 

  • Catalog No:
  • BNC400177-500
  • BNC400177-100
  • Antigen symbol:
  • ICAM3
  • ICAM3
  • Antigen name:
  • CD50
  • CD50
  • Antigen synonyms:
  • ICAMR|Intercellular adhesion molecule 3 (ICAM3)
  • ICAMR|Intercellular adhesion molecule 3 (ICAM3)
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone 101-1D2
  • Clone 101-1D2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 354563
  • 354563
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together