Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CMV-p65 Mouse Monoclonal Antibody (CF594) [clone: CMV100]
 :  Biotium
 
Anti-CMV-p65 Mouse Monoclonal Antibody (CF594) [clone: CMV100]
 :  Biotium

 

  • Catalog No:
  • BNC940100-500
  • BNC940100-100
  • Antigen name:
  • CMV-p65
  • CMV-p65
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone CMV100
  • Clone CMV100
  • Reactivity:
  • Cytomegalovirus
  • Cytomegalovirus
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together