Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-NCAM1 Mouse Monoclonal Antibody (CF640R) [clone: 123C3.D5]
 :  Biotium
 
Anti-NCAM1 Mouse Monoclonal Antibody (CF640R) [clone: 123C3.D5]
 :  Biotium

 

  • Catalog No:
  • BNC400015-500
  • BNC400015-100
  • Antigen symbol:
  • NCAM1
  • NCAM1
  • Antigen name:
  • Neural Cell Adhesion Molecule 1
  • Neural Cell Adhesion Molecule 1
  • Antigen synonyms:
  • NCAM120|NCAM|NCAM180|Leu-19|MSK39|NKH1|NCAM140|Neural Cell Adhesion Molecule
  • NCAM120|NCAM|NCAM180|Leu-19|MSK39|NKH1|NCAM140|Neural Cell Adhesion Molecule
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone 123C3.D5
  • Clone 123C3.D5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 503878
  • 503878
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together