DNA extraction, MPure™ FFPE DNA extraction kit, MP Biomedicals
ICNA117022900
:
- Pk:1 KIT
- Application:
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:FFPE tissues
- Target molecule:DNA
- Description:MPure™ FFPE DNA Extraction Kit, 48 tests
- Environmentally Preferable:
- Description:MPure™ FFPE DNA Extraction Kit, 48 tests
MPure™ FFPE DNA Extraction Kit is used with the MPure-12™ Automated Nucleic Acid Purification System for the extraction and purification of genomic DNA from FFPE (formalin-fixed, paraffin-embedded) tissue samples, providing high quality, high integrity DNA for molecular diagnostics and research use.