Order Entry
Netherlands
ContactUsLinkComponent
Anti-TRAF6 Rabbit Polyclonal Antibody
Anti-TRAF6 Rabbit Polyclonal Antibody
Catalogus # ENZOADIAAP426E
Supplier:  ENZO LIFE SCIENCES
Anti-TRAF6 Rabbit Polyclonal Antibody
Catalogus # ENZOADIAAP426E
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TNF receptor-associated factor 6
  • Antigen symbol:
    TRAF6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Epitope:
    N-Terminal
  • Form:
    Liquid
  • Antigen synonyms:
    TRAF6|Interleukin-1 signal transducer|RNF85|E3 ubiquitin protein ligase|MGC:3310
  • Storage buffer:
    PBS containing 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant mouse TRAF6 (N-terminal fragment)
  • Purification:
    Purified by Immunoaffinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

TRAF proteins are multifunctional signal adaptors that are involved in the activation of the kinase cascades which activate c-Jun N-terminal kinase, p38 mitogen activated protein kinase, and the transcription factor NF-κB.

  • Dilution range: 2 to 4 µg/100 to 500 µl cell extract from 5×106 cells for Immunoprecipitation, 1 µg/ml, ECL for Western blot
  • Detects a band of ~58 kDa by Western blot

Members of the TRAF family form homodimers as well as heterodimers, resulting in extensive diversity and specificity to their function in signal transduction pathways.

Type: Primary
Antigen: TNF Receptor-associated Factor 6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse