Order Entry
Netherlands
ContactUsLinkComponent
Anti-Glutamate decarboxylase 1, Glutamate decarboxylase 2 Mouse Monoclonal Antibody [clone: 9A6]
Anti-Glutamate decarboxylase 1, Glutamate decarboxylase 2 Mouse Monoclonal Antibody [clone: 9A6]
Catalogus # ENZOADIMSA225E
Supplier:  ENZO LIFE SCIENCES
Anti-Glutamate decarboxylase 1, Glutamate decarboxylase 2 Mouse Monoclonal Antibody [clone: 9A6]
Catalogus # ENZOADIMSA225E
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate decarboxylase 1 (brain 67kDa) And glutamate decarboxylase 2
  • Antigen symbol:
    GAD1/GAD2
  • Clonality:
    Monoclonal
  • Clone:
    9A6
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Storage buffer:
    PBS containing 50% glycerol
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant human GAD65
  • Purification:
    Purified by Protein A affinity chromatography
  • Pk:
    100 µG

Specifications

About this item

GAD, glutamic acid decarboxylase, is an integral membrane protein that catalyzes the formation of gamma-aminobutyric acid (GABA), which is an inhibitory neurotransmitter. The biological functions of GAD and GABA extend beyond regulation of neurotransmission to include effects on the immune system and modulation of cell proliferation, protein synthesis, and metabolism.

  • Dilution range: Immunohistochemistry (paraffin sections, 1 - 10 µg/ml), Western blot (1 µg/ml, ECL)
  • Detects bands of ~65kDa and ~67kDa (corresponding to GAD65 and GAD67, respectively) by western blot

Type: Primary
Antigen: Glutamate decarboxylase 1, Glutamate decarboxylase 2
Clonality: Monoclonal
Clone: 9A6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat