


Natriumnitriet 97.0-101.0%, pale yellow powder USP
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7632-00-0
:
Formula:NaNO₂
MDL Number:MFCD00011118
:69 g/mol
UN:1500
ADR:5.1,III
:GHS03
:GHS06
:GHS09
Specification Test Results
- Identity Test:Passes
- Loss on Drying (4 hours over silica):≤0.25%
- Heavy Metals:≤0,002%
- Assay:97,0 - 101,0%
Specification Test Results
Frequently Bought Together