


L(+)-Lactic acid ≥98%, white crystalline powder
:79-33-4
:
:C₃H₆O₃
:MFCD00064266
:90.08 g/mol
:> 100 °C (1000 hPa) (lit.)
:48…58 °C
:1190…1250 kg/m³ (20 °C) (lit.)
:GHS05
:GHS06
:GHS07
:Freezer
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- Melting Point:48 - 58°C
- Optical Rotation:+2.6° ± 2° (c=2.5, water)
- Moisture:≤1%
Frequently Bought Together