Porphobilinogen monohydrate, tan powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:487-90-1
:
Formula:C₁₀H₁₄N₂O₄·H₂O
MDL Number:MFCD00005224
Molecular Weight:244.3 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together