D-(-)-Mannitol ≥99.0%, white powder ACS
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
CAS Number:69-65-8
:
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287
Molecular Weight:182.17 g/mol
Merck Index:13,05769
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥99.0%
- Optical Rotation:+23.3° to +24.3°
- Reducing Sugars:Passes
- Insoluble Matter:≤0.01%
- Loss on Drying:≤0.05%
- Residue After Ignition:≤0.01%
- Titratable Acid:<0.0008 meq/g
- Heavy Metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together