Order Entry
Netherlands
ContactUsLinkComponent
γ-(N-Morpholino)propanesulphonic acid (MOPS), white powder
γ-(N-Morpholino)propanesulphonic acid (MOPS), white powder
  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • UV/Vis: (1M aq soln) OD250 nm:
    ≤0.04
  • UV/Vis: (1M aq soln) OD280 nm:
    ≤0.04
  • pH (10% aq soln):
    3.2 - 4.2
Frequently Bought Together