Gallic acid, fine powder
ICNA0210175080
:
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:250 - 260 °C
: 3,4,5-Trihydroxybenzoic acid
:
:170.12 g/mol
:GHS07
:Ambient
:149-91-7
:C₇H₆O₅
:MFCD00002510
:501
:13,04366