Anti-ADAMTS13 Rabbit Polyclonal Antibody
BSBTPB10028
- Antibody type:Primary
- Antigen name:Adam Metallopeptidase With Thrombospondin Type 1 Motif, 13
- Antigen symbol:ADAMTS13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11093
- Antigen synonyms:C9orf8|vWF-CP|ADAMTS-13|ADAM-TS13|VWFCP
- Size:100 μg
- Pk:100 µG
Rabbit IgG polyclonal antibody for A disintegrin and metalloproteinase with thrombospondin motifs 13(ADAMTS13) detection. Tested with WB, ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: ADAMTS13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat