Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD146 Mouse Monoclonal Antibody (APC-C750) [clone: 47-3D6]
Anti-CD146 Mouse Monoclonal Antibody (APC-C750) [clone: 47-3D6]
  ABNOMAB13810
 :  Abnova
Anti-CD146 Mouse Monoclonal Antibody (APC-C750) [clone: 47-3D6]
  ABNOMAB13810
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD146
  • Clonality:
    Monoclonal
  • Clone:
    47-3D6
  • Conjugation:
    APC (Allophycocyanin)-C750
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Mouse
  • Gene ID:
    929
  • Size:
    100 reactions
  • Pk:
    100 Assays

 

 

Mouse monoclonal antibody raised against human CD14.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: 47-3D6
Conjugation: APC-C750
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,Mouse