Order Entry
Netherlands
ContactUsLinkComponent
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]
Catalog # ABNOMAB13456
Supplier:  Abnova
Anti-ZAP70 Mouse Monoclonal Antibody [clone: 2F3.2]
Catalog # ABNOMAB13456
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ZAP70
  • Clonality:
    Monoclonal
  • Clone:
    2F3.2
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Gene ID:
    7535
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant human ZAP70.

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: 2F3.2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human