Anti-CD34/1171 Mouse Monoclonal Antibody [clone: HPCA1/1171]
ABNOMAB13467
: Abnova
- Antibody type:Primary
- Antigen symbol:CD34/1171
- Clonality:Monoclonal
- Clone:HPCA1/1171
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:947
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human CD34.
Type: Primary
Antigen: CD34/1171
Clonality: Monoclonal
Clone: HPCA1/1171
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human