Anti-TNFRSF8779 Mouse Monoclonal Antibody [clone: Ki-1/779]
Catalog # ABNOMAB13464
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:TNFRSF8779
- Clonality:Monoclonal
- Clone:Ki-1/779
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:943
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human TNFRSF8.
Type: Primary
Antigen: TNFRSF8779
Clonality: Monoclonal
Clone: Ki-1/779
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human