Anti-MLANA/788 Mouse Monoclonal Antibody [clone: MLANA/788]
ABNOMAB13269
: Abnova
- Antibody type:Primary
- Antigen symbol:MLANA/788
- Clonality:Monoclonal
- Clone:MLANA/788
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:2315
- Size:100 μg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human MLANA.
Type: Primary
Antigen: MLANA/788
Clonality: Monoclonal
Clone: MLANA/788
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human