Anti-GYPA280 Mouse Monoclonal Antibody [clone: GYPA/280]
Catalog # ABNOMAB13300
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:GYPA280
- Clonality:Monoclonal
- Clone:GYPA/280
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Epitope:Full-length
- Gene ID:2993
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human GYPA.
Type: Primary
Antigen: GYPA280
Clonality: Monoclonal
Clone: GYPA/280
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human