Anti-Adenovirus Mouse Monoclonal Antibody [clone: J313]
Catalog # ABNOMAB13494
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adenovirus
- Antigen symbol:Adenovirus
- Clonality:Monoclonal
- Clone:J313
- Host:Mouse
- Isotype:IgG1
- Reactivity:Adenovirus
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Mouse monoclonal antibody raised against adenovirus.
Type: Primary
Antigen: Adenovirus
Clonality: Monoclonal
Clone: J313
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Adenovirus