Anti-Streptococcus group B Rabbit Polyclonal Antibody
ABNOPAB29817
: Abnova
- Antibody type:Primary
- Antigen name:Streptococcus group B antigen
- Antigen symbol:Streptococcus group B
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Streptococcus
- Size:500 μg
- Pk:500 µG
Rabbit polyclonal antibody raised against Streptococcus group B.
Type: Primary
Antigen: Streptococcus group B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptococcus