Anti-gB Mouse Monoclonal Antibody [clone: P1123]
Catalog # ABNOMAB13568
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Herpes simplex virus gB
- Antigen symbol:gB
- Clonality:Monoclonal
- Clone:P1123
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Herpes simplex virus
- Size:500 μg
- Pk:500 µG
Specifications
About this item
Mouse monoclonal antibody raised against herpes simplex virus (HSV) gB.
Type: Primary
Antigen: gB
Clonality: Monoclonal
Clone: P1123
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Herpes simplex virus (HSV)