Anti-HSV2 gG3 Mouse Monoclonal Antibody [clone: H2A023]
Catalog # ABNOMAB13589
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Herpes simplex virus type 2 gG3
- Antigen symbol:HSV2 gG3
- Clonality:Monoclonal
- Clone:H2A023
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Herpes simplex virus
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against herpes simplex virus type 2 (HSV-2) gG.
Type: Primary
Antigen: HSV2 gG3
Clonality: Monoclonal
Clone: H2A023
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Herpes simplex virus (HSV)