Anti-KCNQ2 Rabbit Polyclonal Antibody
ABNOPAB29886
: Abnova
- Antibody type:Primary
- Antigen symbol:KCNQ2
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:3785
- Sequence:GNVFATSALRSLRFLQILRMIRMDRRGGTWKLLGSVVYAHSKELVTAWYI
- Size:100 μL
- Pk:100 µl
Rabbit polyclonal antibody raised against partial synthetic protein of human KCNQ2.
Type: Primary
Antigen: KCNQ2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human