Anti-CTPS Rabbit Polyclonal Antibody
Catalog # ABNOPAB30015
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:CTPS
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:1503
- Sequence:SMPFIEAFRQFQFKVKRENFCNIHVSLVPQPSSTGEQKTKPTQNSVRELR
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of human CTPS.
Type: Primary
Antigen: CTPS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human