Order Entry
Netherlands
ContactUsLinkComponent
 
BrightVision Anti-IgG (H+L) (Ultimate Plus kit)
 
BrightVision Anti-IgG (H+L) (Ultimate Plus kit)

 

  • Catalog No:
  • UBVB110HRP
  • UBVB500HRP
  • UBVB999HRP
  • Antigen symbol:
  • IgG (H+L)
  • IgG (H+L)
  • IgG (H+L)
  • Storage temperature:
  • +2 °C to +8 °C
  • +2 °C to +8 °C
  • +2 °C to +8 °C

 

Frequently Bought Together