Specifications
- Antibody type:Primary
- Antigen name:Tumor protein p63
- Antigen symbol:TP63
- Clonality:Monoclonal
- Clone:TP63/11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Seq:03
- Form:Concentrated
- Gene ID:8626
- Antigen synonyms:p51|p73L|OFC8|SHFM4|p63|EEC3|KET|p53CP|p73H|TP73L|NBP|TP53CP|TP53L
- Storage temperature:2°C - 8°C
- Tested applications:Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
- Size:1 ml
- Pk:1 mL
Specifications
About this item
p63 is a homolog of the tumor suppressor p53. It is identified in basal cells in the epithelial layers of a variety of tissues, including epidermis, cervix, urothelium, breast and prostate. It was detected in nuclei of the basal epithelium in normal prostate glands, however, it was not expressed in malignant tumors of the prostate. It has been reported as a useful marker for differentiating benign from malignant lesions in the prostate, particularly when used in combination with markers of high molecular weight cytokeratins and the prostatespecific marker AMACR (P504S). In breast tissue, it has been identified in myoepithelial cells of normal ducts.
- Control: Breast, normal prostate, prostate carcinoma or lung or bladder squamous cell carcinoma
- Dilution Range: 1:400-1:800
Pretreatment: Heat induced epitope retrieval in 10 mM citrate buffer for 20 minutes is required for IHC staining on formalin-fixed, paraffin embedded tissue sections.
Type: Primary
Antigen: TP63
Clonality: Monoclonal
Clone: TP63/11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human