Order Entry
Netherlands
ContactUsLinkComponent
 
Reversed phase HPLC columns, Jupiter® C5
  PHEN00F-4052-E0
Reversed phase HPLC columns, Jupiter® C5
  PHEN00F-4052-E0

 

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    5 µm
  • Pore size:
    300 Å
  • Length:
    150 mm
  • Phase:
    C5
  • Pk:
    1

 

 

Jupiter® HPLC columns offer optimised reversed phase solutions for protein and peptide characterisation and purification.

  • Designed to analyse and purify intact proteins