Order Entry
Netherlands
ContactUsLinkComponent
Reversed phase HPLC columns, Prodigy® ODS-2
Reversed phase HPLC columns, Prodigy® ODS-2
Supplier:  PHENOMENEX
Certificates
Reversed phase HPLC columns, Prodigy® ODS-2
Supplier:  PHENOMENEX
Specifications
  • Catalog No:
  • PHEN00A-3300-E0
  • PHEN00F-3300-E0
  • PHEN00G-3300-E0
  • Carbon load:
  • 18,5%
  • 18,5%
  • 18,5%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • pH range:
  • 2,0 - 9,0
  • 2,0 - 9,0
  • 2,0 - 9,0
  • Surface area:
  • 310 m²/g
  • 310 m²/g
  • 310 m²/g