Order Entry
Netherlands
ContactUsLinkComponent
Reversed phase HPLC columns, Luna® NH₂
Reversed phase HPLC columns, Luna® NH₂
  • Catalog No:
  • PHEN00F-4378-E0
  • PHEN00D-4378-E0
  • PHEN00F-4377-A0
  • PHEN00F-4377-B0
  • Carbon load:
  • 9,5%
  • 9,5%
  • 9,5%
  • 9,5%
  • pH range:
  • 1,5 - 11,0
  • 1,5 - 11,0
  • 1,5 - 11,0
  • 1,5 - 11,0
  • Surface area:
  • 400 m²/g
  • 400 m²/g
  • 400 m²/g
  • 400 m²/g
  • USP code:
  • L8
  • L8
  • L8
  • L8
Frequently Bought Together