Normal phase HPLC columns, Luna® Silica(2)
Supplier: PHENOMENEX
Certificates
About this item
Luna® Silica(2) performance is of ultrapure metal-free silica (99,99% purity) and meticulous care is given to the quality control of surface smoothness, pore structure and pore consistency to ensure particles of uniform structure and enhanced mechanical strength.
- Low percentage of 'fines' from damaged silica leading to lower backpressures and enhanced column performance and lifetimes
- High column bed stability enhanced by particle shape uniformity
Commonly applied for Alprazolam, Hydrocodone bitartrate, Hydrocortisone, Fat Soluble Vitamins, Phthalates, Fatty Acids, Lutein, Lycopene, Estradiol.
Frequently Bought Together