Order Entry
Netherlands
ContactUsLinkComponent
Rack for cuvettes
Rack for cuvettes
  HACHLYW915
 :  Hach
undefined
Rack for cuvettes
  HACHLYW915
 :  Hach

 

  • Description:
    Cuvette rack, 10 mm
  • Pk:
    1

 

 

Rack made of PP, with numbered positions for 2×8 cuvettes of 10 mm pathlength.

  • Numbered positions to aid sample identification
  • Autoclavable at 121 °C