Disposable particle filtering respirators, 9310/9312
Catalog # 111-0033
Supplier: 3M
Specifications
- Type:9312
- Size:U*
- Pk:10
- With valve:+
- Class:FFP1
Specifications
About this item
The foldable disposable FFP1 respirators protect against dusts and mists. They are available with or without Cool Flow™ exhalation valve that helps to breathe easier when working for prolonged periods.
- Unique 3-panel design fits a wide range of face shapes and sizes, accommodates facial movements and makes the respirator easy to talk through
- Concealed adjustable nose-clip is metal detectable
- Coloured straps for easy identification of protection class (yellow for P1)
- Made from 3M™ Advanced Electret Filter Material for effective filtration with low breathing resistance
- Do not contain components made from natural rubber latex
Protection factor: 10× Workplace Exposure Standard.
Certifications: AS/NZS 1716:2012