Napsin is a pepsin-like aspartic proteinase, in the A1 clan of the AA clade of proteinases. There are two closely related napsins, napsin A and napsin B. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. It is expressed as a single chain protein in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.1-5
- Visualization: cytoplasmic
- Control: lung adenocarcinoma, kidney
- Dilution Range: 1:100-1:500
References
1. Jagirdar, J. Arch Pathol Lab Med. 2008; 132:384-96.
2. Dejmek A, et al. Diagn Cytopathol. 2007; 35:493-7.
3. Inamura K, et al. Am J Surg Pathol. 2005; 29:660-5.
4. Bishop JA, et al. Hum Pathol. 2010; 41:20-5.
5. Ye J, et al. Appl Immunohistochem Mol Morphol. 2011; 19:313-317.
Type: Primary
Antigen: NAPSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- ILP3523-C01
- ILP3523-C05
- VWRKILP3523-C1
- Antigen symbol:
- NAPSA
- NAPSA
- NAPSA
- Antigen name:
- Napsin A aspartic peptidase
- Napsin A aspartic peptidase
- Napsin A aspartic peptidase
- Antigen synonyms:
- NAPA|Kdap|KAP|NAP1
- NAPA|Kdap|KAP|NAP1
- NAPA|Kdap|KAP|NAP1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Gene ID:
- 9476
- 9476
- 9476
- Isotype:
- IgG
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C