Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CEACAM5 Rabbit Polyclonal Antibody
 
Anti-CEACAM5 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • ILP83120-C01
  • ILP83120-C05
  • VWRKILP83120-C1
  • Antigen symbol:
  • CEACAM5
  • CEACAM5
  • CEACAM5
  • Antigen name:
  • Carcinoembryonic antigen related 
  • Carcinoembryonic antigen related 
  • Carcinoembryonic antigen related 
  • Antigen synonyms:
  • CEA|CD66e
  • CEA|CD66e
  • CEA|CD66e
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Gene ID:
  • 1048
  • 1048
  • 1048
  • Isotype:
  • IgG
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

 

Frequently Bought Together