
1-Boc-4-(methylamino)piperidine 95%
Catalog # ACRO454800010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥94.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:214.3046 g/mol
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:147539-41-1
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD02259411