Order Entry
Netherlands
ContactUsLinkComponent
 
 
3-Nitro-1H-pyrazole 97%
 
undefined
 
3-Nitro-1H-pyrazole 97%

 

  • Appearance:
    White to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    175 to 178 °C
  • HPLC:
    ≥96.0 %

 

 

Frequently Bought Together