
4'-Fluorobiphenyl-4-sulphonyl chloride 97%
Catalog # ACRO450320050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:80 to 85 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:270.71 g/mol
Melting Pt:82…84 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:116748-66-4
Formula:C₁₂H₈ClFO₂S
MDL Number:MFCD01631921