
5-Amino-3-methyl-1-phenyl-1H-pyrazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1131-18-6
:
Formula:C₁₀H₁₁N₃
MDL Number:MFCD00020727
:173.21 g/mol
Boiling Pt:333 °C (760 mmHg)
Melting Pt:113…115 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to brown Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:114 to 117 °C
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together