Sodium formate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-53-7
:
Formula:HCO₂Na
MDL Number:MFCD00013101
:68.01 g/mol
Boiling Pt:360 °C (760 mmHg)
Melting Pt:258…261 °C
Density:1,919 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration with KMnO4:≥99.0 %
- Chloride (Cl):≤0.001 %
- Sulfate (SO4):≤0.001 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together